Assay Details
| Gene Symbol: tlx3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170766.1 | NP_739572.1 | 1 - 2 | 527 | 94 | |
| GenBank mRNA | AF398519.1 | - | 1 - 2 | 527 | 94 | |
| BC162310.1 | - | 1 - 2 | 469 | 94 | ||
| BC162334.1 | - | 1 - 2 | 469 | 94 | ||
More Information
Set Membership: |
|

tlx3b
Genomic Map