Assay Details
Gene Symbol: adssl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173237.2 | NP_775344.2 | 10 - 11 | 1229 | 72 | |
GenBank mRNA | AY099534.1 | - | 10 - 11 | 1103 | 72 | |
BC051613.1 | - | 10 - 11 | 1089 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
