Assay Details
Gene Symbol: adssl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173237.2 | NP_775344.2 | 5 - 6 | 637 | 96 | |
GenBank mRNA | AY099534.1 | - | 5 - 6 | 511 | 96 | |
BC051613.1 | - | 5 - 6 | 499 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
