Assay Details
| Gene Symbol: scinla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178131.3 | NP_835232.2 | 4 - 5 | 579 | 80 | |
| GenBank mRNA | AF175294.1 | - | 4 - 5 | 597 | 80 | |
| BC094991.1 | - | 4 - 5 | 594 | 80 | ||
| BC116607.1 | - | 4 - 5 | 549 | 80 | ||
| BC125898.1 | - | 4 - 5 | 550 | 80 | ||
| BC134865.1 | - | 4 - 5 | 579 | 80 | ||
| BC155581.1 | - | 4 - 5 | 716 | 80 | ||
More Information
Set Membership: |
|

scinla
Genomic Map