Assay Details
| Gene Symbol: scinla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178131.3 | NP_835232.2 | 8 - 9 | 1045 | 134 | |
| GenBank mRNA | AF175294.1 | - | 8 - 9 | 1063 | 134 | |
| BC116607.1 | - | 8 - 9 | 1015 | 134 | ||
| BC125898.1 | - | 8 - 9 | 1016 | 134 | ||
| BC134865.1 | - | 8 - 9 | 1045 | 134 | ||
| BC155581.1 | - | 8 - 9 | 1182 | 134 | ||
More Information
Set Membership: |
|

scinla
Genomic Map