Assay Details
Gene Symbol: anxa11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181766.2 | NP_861431.1 | 11 - 12 | 1229 | 106 | |
GenBank mRNA | AY178802.2 | - | 11 - 12 | 1229 | 106 | |
BC068366.1 | - | 11 - 12 | 1212 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
