Assay Details
| Gene Symbol: fgf8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182856.1 | NP_878276.1 | 2 - 3 | 564 | 117 | |
| GenBank mRNA | AB100171.1 | - | 2 - 3 | 564 | 117 | |
| BC163345.1 | - | 2 - 3 | 325 | 117 | ||
| BC163362.1 | - | 2 - 3 | 325 | 117 | ||
More Information
Set Membership: |
|

fgf8b
Genomic Map