Assay Details
| Gene Symbol: oaz1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194264.2 | NP_919240.2 | 3 - 4 | 505 | 87 | |
| GenBank mRNA | AB017117.1 | - | 3 - 4 | 487 | 87 | |
| BC056307.1 | - | 3 - 4 | 462 | 87 | ||
| BC067606.1 | - | 3 - 4 | 459 | 87 | ||
| BC158207.1 | - | 3 - 4 | 462 | 87 | ||
| BC163928.1 | - | 3 - 4 | 480 | 87 | ||
| BC163939.1 | - | 3 - 4 | 480 | 87 | ||
| EH453522.1 | - | 3 - 4 | 531 | 87 | ||
| EH612461.1 | - | 2 - 3 | 744 | 87 | ||
More Information
Set Membership: |
|

oaz1a
Genomic Map