Assay Details
| Gene Symbol: hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194368.1 | NP_919349.1 | 9 - 10 | 1226 | 120 | |
| XM_009296811.3 | XP_009295086.1 | 10 - 11 | 1615 | 120 | ||
| XM_021469746.1 | XP_021325421.1 | 9 - 10 | 1222 | 120 | ||
| XM_021469747.1 | XP_021325422.1 | 9 - 10 | 1203 | 120 | ||
| XM_021469748.1 | XP_021325423.1 | 9 - 10 | 1443 | 120 | ||
| XM_021469749.1 | XP_021325424.1 | 9 - 10 | 1470 | 120 | ||
| GenBank mRNA | AF473824.1 | - | 9 - 10 | 1226 | 120 | |
| BC056550.1 | - | 9 - 10 | 1258 | 120 | ||
| BC153623.1 | - | 10 - 11 | 1457 | 120 | ||
More Information
Set Membership: |
|

hnf4a
Genomic Map