Assay Details
| Gene Symbol: arpc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194378.4 | NP_919359.2 | 3 - 4 | 483 | 140 | |
| GenBank mRNA | BC070012.1 | - | 3 - 4 | 408 | 140 | |
| EH437356.1 | - | 3 - 4 | 512 | 140 | ||
More Information
Set Membership: |
|

arpc5a
Genomic Map