Assay Details
Gene Symbol: dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194404.1 | NP_919385.1 | 5 - 6 | 717 | 87 | |
GenBank mRNA | AF286179.1 | - | 5 - 6 | 717 | 87 | |
BC067602.1 | - | 5 - 6 | 729 | 87 | ||
BC092678.1 | - | 5 - 6 | 713 | 87 | ||
BC164850.1 | - | 5 - 6 | 727 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
