Assay Details
| Gene Symbol: mmp14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194414.1 | NP_919395.1 | 4 - 5 | 1045 | 59 | |
| XM_005171310.4 | XP_005171367.1 | 5 - 6 | 973 | 59 | ||
| GenBank mRNA | AY324197.1 | - | 4 - 5 | 1045 | 59 | |
| BC162135.1 | - | 4 - 5 | 937 | 59 | ||
More Information
Set Membership: |
|

mmp14b
Genomic Map