Assay Details
Gene Symbol: ca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199215.2 | NP_954685.2 | 5 - 6 | 566 | 102 | |
GenBank mRNA | BC057412.1 | - | 5 - 6 | 539 | 102 | |
MG797497.1 | - | 5 - 6 | 507 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
