Assay Details
| Gene Symbol: eif4ebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199645.1 | NP_955939.1 | 2 - 3 | 359 | 128 | |
| GenBank mRNA | BC057433.1 | - | 2 - 3 | 359 | 128 | |
| BC154323.1 | - | 2 - 3 | 375 | 128 | ||
| BC155749.1 | - | 2 - 3 | 341 | 128 | ||
More Information
Set Membership: |
|

eif4ebp1
Genomic Map