Assay Details
Gene Symbol: znf207b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199665.2 | NP_955959.1 | 2 - 3 | 293 | 146 | |
XM_009302342.3 | XP_009300617.1 | 2 - 3 | 304 | 146 | ||
XM_021476738.1 | XP_021332413.1 | 2 - 3 | 305 | 146 | ||
GenBank mRNA | AY422996.1 | - | 2 - 3 | 238 | 146 | |
BC049335.1 | - | 2 - 3 | 296 | 146 | ||
BC065453.1 | - | 2 - 3 | 259 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
