Assay Details
| Gene Symbol: hmox1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127516.1 | NP_001120988.1 | 6 - 7 | 941 | 106 | |
| GenBank mRNA | AI722432.1 | - | 1 - 2 | 191 | 106 | |
| BC061954.1 | - | 6 - 7 | 761 | 106 | ||
More Information
Set Membership: |
|

hmox1a
Genomic Map