Assay Details
Gene Symbol: memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199794.1 | NP_956088.1 | 5 - 6 | 706 | 66 | |
GenBank mRNA | BC044360.1 | - | 5 - 6 | 706 | 66 | |
BC155284.1 | - | 5 - 6 | 712 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
