Assay Details
Gene Symbol: bin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199796.1 | NP_956090.1 | 10 - 11 | 960 | 80 | |
XM_005166011.4 | XP_005166068.1 | 9 - 10 | 1082 | 80 | ||
GenBank mRNA | BC044473.1 | - | 10 - 11 | 960 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
