Assay Details
Gene Symbol: bin2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199796.1 | NP_956090.1 | 4 - 5 | 423 | 78 | |
XM_005166011.4 | XP_005166068.1 | 3 - 4 | 545 | 78 | ||
GenBank mRNA | BC044473.1 | - | 4 - 5 | 423 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
