Assay Details
Gene Symbol: tada3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199926.1 | NP_956220.1 | 4 - 5 | 628 | 73 | |
GenBank mRNA | BC055157.1 | - | 4 - 5 | 628 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
