Assay Details
Gene Symbol: tada3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199926.1 | NP_956220.1 | 8 - 9 | 1160 | 59 | |
GenBank mRNA | BC055157.1 | - | 8 - 9 | 1160 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
