Assay Details
Gene Symbol: odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199958.1 | NP_956252.1 | 3 - 4 | 579 | 127 | |
XM_021467596.1 | XP_021323271.1 | 3 - 4 | 633 | 127 | ||
XM_021467597.1 | XP_021323272.1 | 3 - 4 | 997 | 127 | ||
GenBank mRNA | BC054587.1 | - | 3 - 4 | 579 | 127 | |
BC163161.1 | - | 3 - 4 | 404 | 127 | ||
BC163187.1 | - | 3 - 4 | 404 | 127 | ||
BC164454.1 | - | 2 - 3 | 351 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
