Assay Details
| Gene Symbol: cox7a2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199974.2 | NP_956268.1 | 2 - 3 | 311 | 63 | |
| GenBank mRNA | BC055593.1 | - | 2 - 3 | 313 | 63 | |
| BC165164.1 | - | 2 - 3 | 242 | 63 | ||
More Information
Set Membership: |
|

cox7a2l
Genomic Map