Assay Details
| Gene Symbol: napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200031.1 | NP_956325.1 | 3 - 4 | 519 | 68 | |
| GenBank mRNA | BC056836.1 | - | 3 - 4 | 519 | 68 | |
| BC154324.1 | - | 3 - 4 | 451 | 68 | ||
More Information
Set Membership: |
|

napsa
Genomic Map