Assay Details
Gene Symbol: rundc3ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200149.1 | NP_956443.1 | 4 - 5 | 561 | 82 | |
GenBank mRNA | BC044502.1 | - | 4 - 5 | 561 | 82 | |
BC044540.1 | - | 4 - 5 | 561 | 82 | ||
BC155785.1 | - | 3 - 4 | 744 | 82 | ||
BC165804.1 | - | 3 - 4 | 390 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
