Assay Details
| Gene Symbol: washc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200173.1 | NP_956467.1 | 4 - 5 | 353 | 151 | |
| GenBank mRNA | BC045438.1 | - | 4 - 5 | 353 | 151 | |
More Information
Set Membership: |
|

washc3
Genomic Map