Assay Details
| Gene Symbol: kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200209.1 | NP_956503.1 | 3 - 4 | 711 | 89 | |
| GenBank mRNA | BC045989.1 | - | 3 - 4 | 711 | 89 | |
| BC152675.1 | - | 2 - 3 | 1086 | 89 | ||
More Information
Set Membership: |
|

kbtbd4
Genomic Map