Assay Details
| Gene Symbol: gpx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200222.1 | NP_956516.1 | 2 - 3 | 557 | 126 | |
| GenBank mRNA | BC046044.1 | - | 2 - 3 | 557 | 126 | |
More Information
Set Membership: |
|

gpx8
Genomic Map