Assay Details
Gene Symbol: proca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200356.1 | NP_956650.1 | 5 - 6 | 376 | 92 | |
GenBank mRNA | BC053182.1 | - | 5 - 6 | 376 | 92 | |
BC154334.1 | - | 5 - 6 | 365 | 92 | ||
BC165224.1 | - | 3 - 4 | 285 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
