Assay Details
Gene Symbol: proca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200356.1 | NP_956650.1 | 8 - 9 | 754 | 87 | |
GenBank mRNA | BC053182.1 | - | 8 - 9 | 754 | 87 | |
BC154334.1 | - | 8 - 9 | 743 | 87 | ||
BC165224.1 | - | 6 - 7 | 663 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
