Assay Details
Gene Symbol: proca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200356.1 | NP_956650.1 | 9 - 10 | 869 | 70 | |
GenBank mRNA | BC053182.1 | - | 9 - 10 | 869 | 70 | |
BC154334.1 | - | 9 - 10 | 858 | 70 | ||
BC165224.1 | - | 7 - 8 | 778 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
