Assay Details
| Gene Symbol: dnajc5gb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200400.1 | NP_956694.1 | 4 - 5 | 657 | 108 | |
| XM_005158564.4 | XP_005158621.1 | 4 - 5 | 811 | 108 | ||
| XM_005158565.4 | XP_005158622.1 | 4 - 5 | 632 | 108 | ||
| GenBank mRNA | BC054133.1 | - | 4 - 5 | 657 | 108 | |
More Information
Set Membership: |
|

dnajc5gb
Genomic Map