Assay Details
Gene Symbol: znf800b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200436.1 | NP_956730.1 | 4 - 5 | 445 | 119 | |
GenBank mRNA | BC054916.1 | - | 4 - 5 | 445 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
