Assay Details
| Gene Symbol: stambpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200498.1 | NP_956792.1 | 5 - 6 | 786 | 122 | |
| GenBank mRNA | BC046033.1 | - | 4 - 5 | 582 | 122 | |
| BC055512.1 | - | 5 - 6 | 786 | 122 | ||
| BC154224.1 | - | 5 - 6 | 786 | 122 | ||
| BC171484.1 | - | 5 - 6 | 698 | 122 | ||
More Information
Set Membership: |
|

stambpa
Genomic Map