Assay Details
| Gene Symbol: stambpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200498.1 | NP_956792.1 | 9 - 10 | 1218 | 83 | |
| GenBank mRNA | AY398309.1 | - | 9 - 10 | 1220 | 83 | |
| BC046033.1 | - | 8 - 9 | 1014 | 83 | ||
| BC055512.1 | - | 9 - 10 | 1218 | 83 | ||
| BC154224.1 | - | 9 - 10 | 1216 | 83 | ||
| BC171484.1 | - | 9 - 10 | 1130 | 83 | ||
More Information
Set Membership: |
|

stambpa
Genomic Map