Assay Details
| Gene Symbol: rps27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200502.2 | NP_956796.1 | 5 - 6 | 363 | 91 | |
| GenBank mRNA | BC049478.1 | - | 5 - 6 | 364 | 91 | |
| BC055524.1 | - | 5 - 6 | 365 | 91 | ||
| BC164449.1 | - | 4 - 5 | 350 | 91 | ||
More Information
Set Membership: |
|

rps27a
Genomic Map