Assay Details
| Gene Symbol: tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200596.2 | NP_956890.2 | 4 - 5 | 610 | 114 | |
| GenBank mRNA | BC056764.1 | - | 4 - 5 | 610 | 114 | |
More Information
Set Membership: |
|

tmem30c
Genomic Map