Assay Details
Gene Symbol: tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328426.1 | NP_001315355.1 | 6 - 7 | 873 | 74 | |
GenBank mRNA | BC057515.1 | - | 6 - 7 | 657 | 74 | |
BC165159.1 | - | 5 - 6 | 649 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
