Assay Details
| Gene Symbol: ckmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200697.1 | NP_956991.1 | 3 - 4 | 475 | 68 | |
| XM_005165533.4 | XP_005165590.2 | 4 - 5 | 616 | 68 | ||
| XM_009301791.3 | XP_009300066.1 | 4 - 5 | 567 | 68 | ||
| GenBank mRNA | BC059437.1 | - | 3 - 4 | 475 | 68 | |
| BC153620.1 | - | 4 - 5 | 482 | 68 | ||
| BC154617.1 | - | 3 - 4 | 470 | 68 | ||
| BC171364.1 | - | 3 - 4 | 453 | 68 | ||
More Information
Set Membership: |
|

ckmt2b
Genomic Map