Assay Details
| Gene Symbol: ckmt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200697.1 | NP_956991.1 | 7 - 8 | 1045 | 71 | |
| XM_005165533.4 | XP_005165590.2 | 8 - 9 | 1186 | 71 | ||
| XM_009301791.3 | XP_009300066.1 | 8 - 9 | 1137 | 71 | ||
| GenBank mRNA | BC059437.1 | - | 7 - 8 | 1045 | 71 | |
| BC153620.1 | - | 8 - 9 | 1052 | 71 | ||
| BC154617.1 | - | 7 - 8 | 1040 | 71 | ||
| BC171364.1 | - | 7 - 8 | 1023 | 71 | ||
More Information
Set Membership: |
|

ckmt2b
Genomic Map