Assay Details
Gene Symbol: ndufb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200730.1 | NP_957024.1 | 1 - 2 | 223 | 81 | |
GenBank mRNA | BC059507.1 | - | 1 - 2 | 223 | 81 | |
BC071489.1 | - | 1 - 2 | 221 | 81 | ||
BC154202.1 | - | 1 - 2 | 226 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
