Assay Details
| Gene Symbol: ppa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200733.1 | NP_957027.1 | 1 - 2 | 84 | 126 | |
| GenBank mRNA | BC059512.1 | - | 1 - 2 | 84 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppa1a| Gene Symbol: ppa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200733.1 | NP_957027.1 | 1 - 2 | 84 | 126 | |
| GenBank mRNA | BC059512.1 | - | 1 - 2 | 84 | 126 | |
Set Membership: |
|