Assay Details
| Gene Symbol: uqcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200758.2 | NP_957052.1 | 2 - 3 | 342 | 80 | |
| GenBank mRNA | BC059579.1 | - | 2 - 3 | 238 | 80 | |
| EH499435.1 | - | 2 - 3 | 368 | 80 | ||
More Information
Set Membership: |
|

uqcc2
Genomic Map