Assay Details
| Gene Symbol: kat14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002699.3 | NP_001002699.2 | 9 - 10 | 2162 | 63 | |
| GenBank mRNA | EB900813.1 | - | 1 - 2 | 99 | 63 | |
More Information
Set Membership: |
|

kat14
Genomic Map