Assay Details
| Gene Symbol: arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100039.1 | NP_001093509.1 | 1 - 2 | 46 | 81 | |
| XM_005165358.4 | XP_005165415.1 | 1 - 2 | 421 | 81 | ||
More Information
Set Membership: |
|

arid3c
Genomic Map