Assay Details
| Gene Symbol: arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100039.1 | NP_001093509.1 | 6 - 7 | 1444 | 66 | |
| XM_005165358.4 | XP_005165415.1 | 7 - 8 | 1906 | 66 | ||
More Information
Set Membership: |
|

arid3c
Genomic Map