Assay Details
Gene Symbol: igf2bp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114558.2 | NP_001108030.2 | 13 - 14 | 1483 | 128 | |
XM_005167469.4 | XP_005167526.1 | 13 - 14 | 1464 | 128 | ||
XM_009304504.3 | XP_009302779.1 | 13 - 14 | 2230 | 128 | ||
GenBank mRNA | BC155087.1 | - | 5 - 6 | 465 | 128 | |
DQ998838.1 | - | 13 - 14 | 1483 | 128 | ||
EU296401.2 | - | 13 - 14 | 1690 | 128 | ||
FJ418164.1 | - | 13 - 14 | 1690 | 128 | ||
FJ418165.1 | - | 12 - 13 | 1386 | 128 | ||
FJ418166.1 | - | 12 - 13 | 1386 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
