Assay Details
| Gene Symbol: plvapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114577.1 | NP_001108049.1 | 3 - 4 | 1191 | 108 | |
| GenBank mRNA | BC158204.1 | - | 3 - 4 | 1191 | 108 | |
More Information
Set Membership: |
|

plvapa
Genomic Map