Assay Details
Gene Symbol: abca1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114586.2 | NP_001108058.2 | 3 - 4 | 654 | 148 | |
XM_005173080.4 | XP_005173137.1 | 4 - 5 | 463 | 148 | ||
GenBank mRNA | BC153976.1 | - | 3 - 4 | 655 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
