Assay Details
| Gene Symbol: gabrz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114742.2 | NP_001108214.2 | 3 - 4 | 373 | 132 | |
| GenBank mRNA | BC159112.1 | - | 3 - 4 | 373 | 132 | |
More Information
Set Membership: |
|

gabrz
Genomic Map